Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Aluminum wire, 0.127mm (0.005 in.) dia., hard, Puratronic™, 99.999% (metals basis)

Click to view available options
Quantity:
25m
5m
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
7429-90-5
Al
26.982
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium,metal,powder,aluminio,metana,adom,alumina fibre,aluminium flake,dust,noral aluminium
5359268
CHEBI:33629
aluminum
[Al]
Quantity 5m
Weight ≈0.034g/m
Odor Odorless
Physical Form Wire, ≈0.034g/m,Temper: as drawn
Percent Purity 99.999%
Grade Puratronic™
Chemical Name or Material Aluminum wire, 0.127mm (0.005 in.) dia., Hard, Temper: as drawn
Assay (metals basis)

EINECSNumber : 231-072-3

TSCA : Yes

Recommended Storage : Ambient temperatures