Alfa Aesar™ Aluminum wire, 0.81mm (0.032 in.) dia., annealed, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 43336H7

Quantity 250m

 View more versions of this product

Catalog No. AA43336H7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    MDL Number MFCD00134029
    Chemical Name or Material Aluminum wire, 0.81mm (0.032 in.) dia., Annealed, Temper: soft
    Grade Puratronic™

    Specifications

    CAS 7429-90-5
    Physical Form Wire
    Odor Odorless
    Weight ≈1.4g/m
    Quantity 250m
    TSCA TSCA
    MDL Number MFCD00134029
    Chemical Name or Material Aluminum wire, 0.81mm (0.032 in.) dia., Annealed, Temper: soft
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit