Aluminum Wire, 1.5mm (0.059 in.) dia, annealed, Puratronic™, 99.999% (Metals basis), Alfa Aesar™

Form: Approx. 4.8g/m

Manufacturer: Alfa Aesar 10765G1

Quantity 1m

 View more versions of this product

Catalog No. AA10765G1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7429-90-5
    Physical Form Wire, 1.5mm (0.06 in.) dia.
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00134029
    Chemical Name or Material Aluminum, Annealed

    Specifications

    Quantity 1m
    Color Silver
    Physical Form Wire, 1.5mm (0.06 in.) dia.
    Assay Percent Range 99.999% (Metals basis)
    MDL Number MFCD00134029
    CAS 7429-90-5
    Odor Odorless
    Appearance Silver-colored wire
    Chemical Name or Material Aluminum, Annealed

    SDS & Safety Documents

    • Alfa Aesar and Acros Organics™ truly are better together: You'll receive $150 of Alfa Aesar or Acros Organics products when you buy $300 of Alfa Aesar or Acros Organics products.

      Buy Now

      Expires 2017-01-30 Redemption Only!

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit