Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALX1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548910

Catalog No. PA548910

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Transcriptional activator that acts at a palindromic recognition sequence to enhance the activity of the SV40 and TK promoters. Functions as a repressor with the prolactin promoter in vivo. May play a role in chondrocyte differentiation and may also influence cervix development.
Specifications

Specifications

ALX1
Polyclonal
PBS with 0.09% sodium azide
Q15699
ALX homeobox protein 1, Cartilage homeoprotein 1, CART-1, ALX1, CART1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8092
Western Blot
Unconjugated
ALX1
Liquid
ALX1
KLH conjugated synthetic peptide between 97-122 amino acids from the Central region of human ALX1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit