Learn More
ALX3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP18028520UL
 View more versions of this product
Catalog No. NBP18028520
Add to cart
Description
Description
ALX3 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications
Specifications
ALX3
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
ALX3
Synthetic peptide directed towards the N terminal of mouse ALX3 (NP_031467). Peptide sequence MDPERCAPFSVGPAAGPYAAAGDEAPGPQGTPDAAPHLHPAPPRGPRLSR.
37 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Western Blot
Western Blot 2.5 ug/ml
O70137
ALX homeobox 3, aristaless-like homeobox 3, homeobox protein aristaless-like 3, MGC138212, MGC141988, Proline-rich transcription factor ALX3
Rabbit
IgG
Protein A purified
RUO
Primary
257