Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AM 114, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
856849-35-9
C20H21B2NO5
377.01
SRPIKXGUPAKTIZ-UHFFFAOYSA-N
3, 5-Bis-[benzylidene-4-boronic acid]-1-methylpiperidin-4-one
57369225
[4-[[5-[(4-boronophenyl)methylidene]-1-methyl-4-oxopiperidin-3-ylidene]methyl]phenyl]boronic acid
B(C1=CC=C(C=C1)C=C2CN(CC(=CC3=CC=C(C=C3)B(O)O)C2=O)C)(O)O
This item is not returnable. View return policy
856849-35-9
C20H21B2NO5
377.01
SRPIKXGUPAKTIZ-UHFFFAOYSA-N
3, 5-Bis-[benzylidene-4-boronic acid]-1-methylpiperidin-4-one
57369225
[4-[[5-[(4-boronophenyl)methylidene]-1-methyl-4-oxopiperidin-3-ylidene]methyl]phenyl]boronic acid
B(C1=CC=C(C=C1)C=C2CN(CC(=CC3=CC=C(C=C3)B(O)O)C2=O)C)(O)O

20S proteasome inhibitor

Inhibitor of the chymotrypsin-like activity of the 20S proteasome (IC 50 ∽ 1 μM). Displays anticancer activity; inhibits cell growth in human colon cancer HCT116 p53+/+ cells (IC 50 = 1.49 μM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 377.01
Percent Purity >97%
Chemical Name or Material AM 114

Recommended Storage : Store at +4°C