AM 404 (in Tocrisolve 100), Tocris

Anandamide transport inhibitor (in water-soluble emulsion)

Manufacturer: R&D Systems 1685/5

Quantity 5mg

 View more versions of this product

Catalog No. 16-855

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C26H37NO2
    Formula Weight 395.58
    Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
    Chemical Name or Material AM 404 (in Tocrisolve™ 100)

    Specifications

    Formula Weight 395.58
    Quantity 5mg
    Synonym N-(4-Hydroxyphenyl)-5Z,8Z,11Z,14Z-eicosatetraenamide
    Assay Percent Range >98%
    Molecular Formula C26H37NO2
    CAS 198022-70-7
    Recommended Storage Store at +4°C
    Chemical Name or Material AM 404 (in Tocrisolve™ 100)

    Store at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit