Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AM 630, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
164178-33-0
C23H25IN2O3
504.368
JHOTYHDSLIUKCJ-UHFFFAOYSA-N
iodopravadoline,6-iodo-2-methyl-1-2-morpholinoethyl-1h-indol-3-yl 4-methoxyphenyl methanone,6-iodopravadoline,unii-u1lnj6nbka,u1lnj6nbka,6-iodo-2-methyl-1-2-morpholin-4-ylethyl indol-3-yl-4-methoxyphenyl methanone,1-2-morpholin-4-yl ethyl-2-methyl-3-4-methoxybenzoyl-6-iodoindole,6-iodo-3-4-methoxybenzoyl-2-methyl-1-2-morpholin-4-yl ethyl indole,tocris-1120
4302963
[6-iodo-2-methyl-1-(2-morpholin-4-ylethyl)indol-3-yl]-(4-methoxyphenyl)methanone
CC1=C(C2=C(N1CCN3CCOCC3)C=C(C=C2)I)C(=O)C4=CC=C(C=C4)OC
This item is not returnable. View return policy
164178-33-0
C23H25IN2O3
504.368
JHOTYHDSLIUKCJ-UHFFFAOYSA-N
iodopravadoline,6-iodo-2-methyl-1-2-morpholinoethyl-1h-indol-3-yl 4-methoxyphenyl methanone,6-iodopravadoline,unii-u1lnj6nbka,u1lnj6nbka,6-iodo-2-methyl-1-2-morpholin-4-ylethyl indol-3-yl-4-methoxyphenyl methanone,1-2-morpholin-4-yl ethyl-2-methyl-3-4-methoxybenzoyl-6-iodoindole,6-iodo-3-4-methoxybenzoyl-2-methyl-1-2-morpholin-4-yl ethyl indole,tocris-1120
4302963
[6-iodo-2-methyl-1-(2-morpholin-4-ylethyl)indol-3-yl]-(4-methoxyphenyl)methanone
CC1=C(C2=C(N1CCN3CCOCC3)C=C(C=C2)I)C(=O)C4=CC=C(C=C4)OC

Selective CB2 inverse agonist

CB2 inverse agonist (Ki = 31.2 nM) that displays 165-fold selectivity over CB1 receptors. Behaves as a weak partial/inverse agonist at CB1 receptors.
Quantity 10mg
Formula Weight 504.37
Percent Purity >98%
Chemical Name or Material AM 630

Recommended Storage : Desiccate at +4°C