Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AM-Tag Recombinant Mouse Monoclonal Antibody, AbFlex®, Active Motif™

Mouse Recombinant Monoclonal Antibody
Supplier: Active Motif 91111

Description
The AM-Tag antibody is specifically designed for use with Active Motif's unique AM-Tag sequence in combination with the Tag-ChIP-IT® Kit (Catalog No. 53022). Simply clone your protein of interest into Active Motif's pAM_1C Empty Vector (Catalog No. 53023), or add the AM-Tag sequence to the C-terminus of your protein expression vector. Following transfection, cell lysates are prepared and screened by Western blot for expression of the AM-Tag fusion protein using the AM-Tag antibody. Optimization may be required to identify the optimal conditions for transfection and protein expression.Specifications
AM-Tag | |
Monoclonal | |
Unconjugated | |
Mouse | |
Protein A | |
RUO | |
Human | |
Antibody | |
IgG2a |
ChIP assay, Western Blot | |
1 mg/mL | |
200mM HEPES, 50mM sodium acetate with 100mM NaCl, 30% glycerol and 0.035% sodium azide; pH 7.5 | |
This antibody was raised against Active Motif's unique AM-Tag sequence. | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title