Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AM-Tag Mouse anti-Human, Active Motif™ Non-distribution product as customer accommodation.

Mouse Recombinant Monoclonal Antibody

Specifications

Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Antigen AM-Tag
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3708 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91112
10 μg N/A
50-199-3707 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
91111
100 μg N/A
Description

Description

The AM-Tag antibody is specifically designed for use with Active Motif's unique AM-Tag sequence in combination with the Tag-ChIP-IT® Kit (Catalog No. 53022). Simply clone your protein of interest into Active Motif's pAM_1C Empty Vector (Catalog No. 53023), or add the AM-Tag sequence to the C-terminus of your protein expression vector. Following transfection, cell lysates are prepared and screened by Western blot for expression of the AM-Tag fusion protein using the AM-Tag antibody. Optimization may be required to identify the optimal conditions for transfection and protein expression.
Specifications

Specifications

Monoclonal
Primary
AM-Tag
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit