Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR Mouse anti-Human, Clone: AMACR/1723, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14791

Catalog No. 89-034-874


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human AMACR.

  • High specificity
  • High reactivity
Specifications

Specifications

AMACR
AMACR/1723
Mouse monoclonal antibody raised against full length recombinant human AMACR.
In 10 mM PBS.
Liquid
AMACR
Recombinant protein corresponding to full length human AMACR.
Protein A/G purification
RUO
Primary
23600
Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Q9UHK6
RACE
Mouse
IgG
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)