Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR (P504S) Rabbit anti-Human, Clone: 13H4, Biologo

Rabbit Monoclonal Antibody

Manufacturer:  Exalpha Biologicals IncSupplier Diversity Partner P504S05

Encompass

 View more versions of this product

Catalog No. 50-210-1682


Add to cart

Description

Description

AMACR (P504S) Monoclonal antibody specifically detects AMACR (P504S) in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Specifications

Specifications

AMACR (P504S)
Monoclonal
40 μg/mL
PBS with 0.5% BSA and 0.09% sodium azide; pH
Q9UHK6
2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase; Amacr; AMACRD; amcr; CBAS4; Da1-8; Macr1; Marc1; OTTHUMP00000219978; OTTHUMP00000219979; RACE; RM
Human recombinant AMACR (P504S) protein, whole sequence
Ion-exchange chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
13H4
Unconjugated
AMACR
Liquid
Rabbit
IgG
0.5 mL
4° C
Primary
23600
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit