Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMACR (P504S) Rabbit anti-Human, Clone: 13H4, Biologo Non-distribution product as customer accommodation.

Rabbit Monoclonal Antibody

$738.10 - $1433.85

Specifications

Antigen AMACR (P504S)
Clone 13H4
Concentration 40 μg/mL
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-1682 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
P504S05
0.5 mL
Each for $738.10
Add to cart
 
50-210-1683 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
P504S10
1 mL
Each for $1,433.85
Add to cart
 
Description

Description

AMACR (P504S) Monoclonal antibody specifically detects AMACR (P504S) in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Specifications

Specifications

AMACR (P504S)
40 μg/mL
Monoclonal
Liquid
RUO
PBS with 0.5% BSA and 0.09% sodium azide; pH
2-arylpropionyl-CoA epimerase; 2-methylacyl-CoA racemase; Alpha-methylacyl-CoA racemase; alpha-methylacyl-Coenzyme A racemase; Amacr; AMACRD; amcr; CBAS4; Da1-8; Macr1; Marc1; OTTHUMP00000219978; OTTHUMP00000219979; RACE; RM
Human recombinant AMACR (P504S) protein, whole sequence
Monoclonal
Ion-exchange chromatography
Antibody
13H4
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q9UHK6
23600
IgG
Primary
4° C
AMACR
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit