Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amantadine Hydrochloride, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
665-66-7
C10H18ClN
187.71
WOLHOYHSEKDWQH-UHFFFAOYSA-N
hydrogen adamantan-1-amine chloride
[H+].[Cl-].NC12CC3CC(CC(C3)C1)C2
665-66-7
C10H18ClN
187.71
WOLHOYHSEKDWQH-UHFFFAOYSA-N
hydrogen adamantan-1-amine chloride
[H+].[Cl-].NC12CC3CC(CC(C3)C1)C2

AM102, 665-66-7, C10H17N•HCl

Spectrum™ Amantadine Hydrochloride, USP is an antiviral and an antiparkinsonian drug. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 1 kg
CAS Max % 100%
Percent Purity 98.5 to 101.5%