Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps

For light sensitive solvents, chemicals, and samples. Fisherbrand™ Amber Boston Round Bottles with White Polypropylene Pulp/Vinyl Lined Caps are general use bottles for long term storage of UV sensitive contents.

 View more versions of this product

Catalog No. 02-911-746A

  Check Availability
Add to Cart

Includes: Polypropylene Pulp/Vinyl lined cap

    Specifications

    Capacity (English) 2 oz.
    Closure Color White
    Closure Material Polypropylene
    Color Amber
    Material Glass
    Mouth Narrow
    Shape Round
    Capacity (Metric) 60mL
    Product Type Boston Round Bottle
    Closure Type Screw
    Liner Material Pulp/Vinyl
    Neck Finish 20-400
    Quantity 24/Cs.

    Amber Boston Round Bottles
    • Boston round bottles features a narrow screw neck opening, curved shoulders, and a round body
    • Glass offers minimal chemical interference with contents
    • Amber glass bottles absorb all wavelengths in the ultraviolet spectrum
    White Polypropylene Pulp/Vinyl Lined Caps
    • Good impact strength, thermal resistance and pliability
    • Autoclavable
    • Liner is made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit