Learn More
DWK Life Sciences Wheaton™ Amber Glass Jug, 3840mL/130 fl. oz.
Jug is made of amber-tinted Type III Glass.
Manufacturer:  DWK Life Sciences W220982
 View more versions of this product
Catalog No. 03-084-166
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Type III Soda-Lime tinted amber to provide UV protection for light-sensitive materials
  • Available with black phenolic PTFE/PE lined cap, black phenolic PE cone-lined cap, black phenolic rubber lined cap or white PP/PV lined cap.
Specifications
Specifications
3840mL
Phenolic
Amber
6/Cs
Black
130 f. oz.
Rubber
Type III Borosilicate Glass
Glass Jug