Learn More
Qorpak™ Amber Molded Glass Type I Borosilicate Serum Vials
Description
- 100% inspected in clean room environment
- Vials are washed with water for injection and dried in an ISO8 environment
- Vials are depyrogenated in a tunnel according to European and US pharmaccopeias
- Vials are placed in trays for packaging in ISO5 area with ISO7 surrounding
- Sealed with a Tyvek™ lid in ISO5 environment, packed in steribags and placed into abox allowing for sterilization in ISO8 surrounding
- Sterilization is made through ethylene oxide and a process validated according to the ISO11135 normal
Specifications
Specifications
| Color | Amber |
| Diameter (Metric) Outer | 32 mm |
| Material | Borosilicate Glass |
| Pre-slit | No |
| Sterility | Sterile |
| Disposable | Single-use |
| Volume (Metric) | 20 mL |
| For Use With (Application) | Lyophilization |
| Graduated | No |
| No. per Case | 480/Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.