Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Qorpak™ Amber Molded Glass Type I Borosilicate Serum Vials
Click to view available options
Volume (Metric):
10 mL
20 mL
50 mL
100 mL
Description
- 100% inspected in clean room environment
- Vials are washed with water for injection and dried in an ISO8 environment
- Vials are depyrogenated in a tunnel according to European and US pharmaccopeias
- Vials are placed in trays for packaging in ISO5 area with ISO7 surrounding
- Sealed with a Tyvek™ lid in ISO5 environment, packed in steribags and placed into abox allowing for sterilization in ISO8 surrounding
- Sterilization is made through ethylene oxide and a process validated according to the ISO11135 normal
Specifications
Specifications
Color | Amber |
Diameter (Metric) Outer | 25.4 mm |
Material | Borosilicate Glass |
Pre-slit | No |
Sterility | Sterile |
Disposable | Single-use |
Volume (Metric) | 10 mL |
For Use With (Application) | Lyophilization |
Graduated | No |
No. per Case | 640/Cs. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction