Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Please call 1-800-766-7000 to place your order or try our site again later.
Primeline Industries Amber Thin-Wall Tubing
Noncontaminating; free of sulfur and talc
.
$38.50 - $54.72
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Price | Quantity & Availability | |||||
S50637
![]() ![]() |
Primeline Industries
034RA |
6.4 mm |
Pack for $50.30
|
N/A |
S50636
![]() ![]() |
Primeline Industries
026RA |
4.8 mm |
Pack for $38.50
|
N/A |
S50638
![]() ![]() |
Primeline Industries
038RA |
7.9 mm |
Pack for $54.72
|
N/A |
Description
High-quality seamless tubing produced by dipping mandrel into natural latex.
- Thickness: 1.6mm (0.06 in.)
- Soft, pliable and non-tacky
- Withstands autoclaving
- Length: 50 ft. (15.2m)
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title