Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ CG-50 - type 1, ion-exchange resin, Thermo Scientific™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
9042-11-9
MFCD00132693
9042-11-9
MFCD00132693
Ion exchange resin
Packaging Plastic bottle
Fieser 01,511; 04,266; 05,355; 06,302
Merck Index 14, 382
Quantity 25g
Chemical Name or Material Amberlite™ CG-50 - type 1

Recommended Storage : Indoors +20°C

RUO – Research Use Only