Alfa Aesar™ Amberlite™ FPC3500

Manufacturer: Alfa Aesar™ H331010B

Quantity 1000g

 View more versions of this product

Catalog No. AAH331010B

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD08460528
    Chemical Name or Material Amberlite™ FPC3500

    Specifications

    MDL Number MFCD08460528
    Quantity 1000g
    Chemical Name or Material Amberlite™ FPC3500

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit