Learn More
Alfa Aesar™ Amberlite™ FPC3500(H), ion exchange resin
Manufacturer:  Alfa Aesar™ 45903A1
Quantity 1kg
 View more versions of this product
Catalog No. AA45903A1
  Check Availability
Add to cart
Specifications
Description
  • Suitable for the neutralization of strong bases, the recovery of metallic ions, and the isolation and concentration of antibiotics, basic amino acids, enzymes, and peptides
Description & Specifications
Chemical Identifiers
MFCD08460528
14,832
Amberlite™ FPC3500 (H) ion exchange resin
Specifications
1.2g/mL
White
1kg
MFCD08460528
14,832
Amberlite™ FPC3500 (H) ion exchange resin