Amberlite™ IRA-400(Cl), ion exchange resin, Alfa Aesar™

Manufacturer: Alfa Aesar 1724636

Quantity 500g

 View more versions of this product

Catalog No. AA1724636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9002-24-8
    Molecular Formula Styrene-DVB; Strongly basic gel type resin
    MDL Number MFCD00132708
    Chemical Name or Material Amberlite IRA-400, ion exchange resin

    Specifications

    Quantity 500g
    Molecular Formula Styrene-DVB; Strongly basic gel type resin
    CAS 9002-24-8
    MDL Number MFCD00132708
    Chemical Name or Material Amberlite IRA-400, ion exchange resin

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit