Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRA-402, Cl-form, ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
52439-77-7
52439-77-7
Ion exchange resin
Moisture Holding Capacity (Cl): 49.0 to 60.0%
Particle Size Particle size analysis: uniformity coefficient: <1.60, harmonic mean size: 0.600 to 0.750mm,% <0.300mm: <1.0%,%>1.180mm: <5.0%
Packaging Plastic bottle
Volume Total Capacity (Cl):> 1.20 eq/L
Color Yellow
Solubility Information Solubility in water: insoluble.
Quantity 250g
Physical Form Beads
Chemical Name or Material Amberlite™ IRA-402