Alfa Aesar™ Amberlite IRA-402(Cl), ion exchange resin

Manufacturer: Alfa Aesar™ B255560E

Quantity 2500g

 View more versions of this product

Catalog No. AAB255560E

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52439-77-7
    MDL Number MFCD01864002
    Chemical Name or Material Amberlite IRA-402(Cl), ion exchange resin

    Specifications

    TSCA TSCA
    MDL Number MFCD01864002
    CAS 52439-77-7
    Quantity 2500g
    Chemical Name or Material Amberlite IRA-402(Cl), ion exchange resin

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit