Alfa Aesar™ Amberlite IRA-402(OH), ion exchange resin

Manufacturer: Alfa Aesar™ A1818522

Quantity 100g

 View more versions of this product

Catalog No. AAA1818522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 52439-77-7
    MDL Number MFCD00145564
    Merck Index 14,382
    Chemical Name or Material Amberlite IRA-402(OH), ion exchange resin

    Specifications

    Quantity 100g
    MDL Number MFCD00145564
    CAS 52439-77-7
    Merck Index 14,382
    Chemical Name or Material Amberlite IRA-402(OH), ion exchange resin

    Description

    Applications
    Amberlite IRA-402(OH) ion exchange resin is a strongly basic gel-type anion exchange resin. It is used to adsorb uranium(VI) from sulphate, removal of cyanide and zinc cyanide. It is also used to produce perchlorate and the free nucleoside. Further, it is used for water conditioning.

    Solubility
    Insoluble in water.

    Notes
    Incompatible with strong oxidizing agents, strong bases and acids.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit