Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRA-900 Cl ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
9050-97-9
C4H12ClN
109.60
MFCD00011628
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
6379
CHEBI:7070
tetramethylazanium;chloride
[Cl-].C[N+](C)(C)C
9050-97-9
C4H12ClN
109.60
MFCD00011628
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl
6379
CHEBI:7070
tetramethylazanium;chloride
[Cl-].C[N+](C)(C)C
Ion exchange resin
Moisture Holding Capacity (Cl-): 58 to 64%
Particle Size Particle size analysis : uniformity coefficient: <1.8, harmonic mean size: 0.650 to 0.820mm, <0.300mm: <0.5%,> 1.180mm: <3.0%
Packaging Plastic bottle
Volume Total Capacity (Cl-):> 1.0 eq/L
Color Beige-Brown to Ivory
Fieser 01,511; 02,267; 04,267; 06,302; 07,182
Quantity 100g
Physical Form Beads
Chemical Name or Material Amberlite™ IRA-900