Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite™ IRA-958(Cl), ion exchange resin, Thermo Scientific Chemicals

Removes Color, organics, and Cyanide

$123.00 - $358.00

Specifications

Chemical Name or Material Amberlite IRA-958(Cl)
Physical Form Beads
MDL Number MFCD00211600
Color White
Solubility Information Insoluble in water.
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AA42702A1
View Documents
Thermo Scientific Chemicals
042702A1
1 kg
Each for $123.00
Order in Bulk

Request bulk or custom formats

Add to Cart
 
AA42702A7
View Documents
Thermo Scientific Chemicals
042702A7
5 kg
Each for $358.00
Order in Bulk

Request bulk or custom formats

Add to Cart
 
AA4270230
View Documents
Thermo Scientific Chemicals
04270230
250 g Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

Removal of color, organics, and cyanideAmberlite™ IRA-958(Cl), ion exchange resin is used as an organic scavenger for the adsorption of natural organic matter from surface water. It is an acrylic resin for decolorization and the removal of organic material. Further, it is useful for decolorizing sugar juices and cane syrups, citric acid purification and capturing metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Amberlite IRA-958(Cl)
MFCD00211600
Insoluble in water.
Beads
White
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit