Alfa Aesar™ Amberlite™ IRC-76, ion exchange resin

Manufacturer: Alfa Aesar™ 3938436

Quantity 500g

 View more versions of this product

Catalog No. AA3938436

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 11098-83-2
    MDL Number MFCD00145802
    Merck Index 14,382
    Chemical Name or Material Amberlite™ IRC-76, Ion exchange resin, Acrylic-DVB; Weakly acidic gel type cation resin

    Specifications

    MDL Number MFCD00145802
    Quantity 500g
    Odor Odorless
    CAS 11098-83-2
    Merck Index 14,382
    Chemical Name or Material Amberlite™ IRC-76, Ion exchange resin, Acrylic-DVB; Weakly acidic gel type cation resin

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit