Learn More
MilliporeSigma™ Supelco™ AmberLite MB20 H/OH Mixed Bed Ion Exchange Resin, Hydrogen and Hydroxide Form, 20-50 mesh
Formerly Dowex™ Marathon™ MR-3 H/OH
Supplier:  MilliporeSigma™ Supelco™ 428736500G
 View more versions of this product
Catalog No. 11-100-2509
Add to Cart
Description
Description
Dowex Marathon MR-3 is a premium, premixed, mixed bed product designed for polishing demineralized water to conductivities < 0.1 muS/cm. Marathon MR-3 is a 1:1 equivalent mixture of Marathon C-10 (H) and Marathon A (OH)
Specifications
Specifications
20/50 Mesh
0 to 14
Marathon
1.0 meq/mL by wetted bed volume (anion)
Poly Bottle
500 g
HCR-S[H+] and SBR[OH-]: in accordance
MFCD00166706
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow