Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ AmberLite MB20 H/OH Mixed Bed Ion Exchange Resin, Hydrogen and Hydroxide Form, 20-50 mesh
Formerly Dowex™ Marathon™ MR-3 H/OH
$110.00 - $174.00
Specifications
Particle Size | 20/50 Mesh |
---|---|
Packaging | Poly Bottle |
pH | 0 to 14 |
Product Line | Marathon |
Description | HCR-S[H+] and SBR[OH-]: in accordance |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-100-2509
|
MilliporeSigma™ Supelco™
428736500G |
500 g |
Each for $110.00
|
|
|||||
11-100-2508
|
MilliporeSigma™ Supelco™
4287361KG |
1 kg |
Each for $174.00
|
|
|||||
11-100-2510
|
MilliporeSigma™ Supelco™
428736100G |
100 g | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Dowex Marathon MR-3 is a premium, premixed, mixed bed product designed for polishing demineralized water to conductivities < 0.1 muS/cm. Marathon MR-3 is a 1:1 equivalent mixture of Marathon C-10 (H) and Marathon A (OH)Specifications
20/50 Mesh | |
0 to 14 | |
HCR-S[H+] and SBR[OH-]: in accordance | |
MFCD00166706 |
Poly Bottle | |
Marathon | |
1.0 meq/mL by wetted bed volume (anion) |
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title