Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlite(R), IR-120 H, 16-50 Mesh, Spectrum™

Click to view available options
Quantity:
500 g
6 x 500 g
Packaging:
Amber Glass Bottle
39389-20-3
C18H18O3S
314.40
SIWVGXQOXWGJCI-UHFFFAOYSA-N
1,2-diethenylbenzene; 2-ethenylbenzene-1-sulfonic acid
C=CC1=CC=CC=C1C=C.OS(=O)(=O)C1=CC=CC=C1C=C
39389-20-3
C18H18O3S
314.40
SIWVGXQOXWGJCI-UHFFFAOYSA-N
1,2-diethenylbenzene; 2-ethenylbenzene-1-sulfonic acid
C=CC1=CC=CC=C1C=C.OS(=O)(=O)C1=CC=CC=C1C=C

AMB40, 39389-20-3

Spectrum™ Amberlite(R), IR-120 H, 16-50 Mesh
CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 6 x 500 g