Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlyst™ 15, (dry) ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Plastic bottle
9037-24-5
138.992
MFCD00145841
YZUPZGFPHUVJKC-UHFFFAOYSA-N
80972
COCCBr
9037-24-5
138.992
MFCD00145841
YZUPZGFPHUVJKC-UHFFFAOYSA-N
80972
COCCBr
Ion exchange resin
Additional Information weight total capacity (H):> 4.70 eq/kg
Particle Size Particle size analysis:% <0.300mm: <1.0%
Packaging Plastic bottle
Water 1.5% max.
Fieser 05,355; 06,302; 09,256
Merck Index 15, 377
Quantity 500g
Physical Form Beads
Chemical Name or Material Amberlyst™ 15