Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amberlyst™ 15(wet) ion-exchange resin, ACROS Organics™

Click to view available options
Quantity:
25g
500g
5kg
Packaging:
Plastic bottle
Plastic drum
9037-24-5
C3H7BrO
138.992
MFCD00145841
YZUPZGFPHUVJKC-UHFFFAOYSA-N
2-bromoethyl methyl ether,2-methoxyethyl bromide,ethane, 1-bromo-2-methoxy,ether, 2-bromoethyl methyl,methoxyethyl bromide,2-bromoethylmethylether,bromoethyl methyl ether,2-bromoethylmethyl ether,2-bromoethylmethyl-ether,2-bromo-1-methoxyethane
80972
1-bromo-2-methoxyethane
COCCBr
9037-24-5
C3H7BrO
138.992
MFCD00145841
YZUPZGFPHUVJKC-UHFFFAOYSA-N
2-bromoethyl methyl ether,2-methoxyethyl bromide,ethane, 1-bromo-2-methoxy,ether, 2-bromoethyl methyl,methoxyethyl bromide,2-bromoethylmethylether,bromoethyl methyl ether,2-bromoethylmethyl ether,2-bromoethylmethyl-ether,2-bromo-1-methoxyethane
80972
1-bromo-2-methoxyethane
COCCBr
Ion exchange resin
Additional Information weight total capacity (H):> 4.70 eq/kg
Moisture Holding Capacity (H): 52.0 to 57.0%
Particle Size Particle size analysis: uniformity coefficient: <1.70, harmonic mean size: 0.600 to 0.850mm,% <0.355mm: < 1.0%,%>1.180mm: <5.0%
Packaging Plastic drum
Volume Total Capacity (H):> 1.70 eq/L
Fieser 05,355; 06,302; 09,256
Color Brown-Gray
Quantity 5kg
Physical Form Beads
Chemical Name or Material Amberlyst™ 15