Learn More
Invitrogen Barrier (Filter) Tips, 100 µL size
Aerosol filter prevents cross-contamination of samples
Manufacturer: Invitrogen AM12648
Description
Compatible with Eppendorf, Finnpipette, Oxford Benchmate, Pipetman, Proline, Rainin, Socorex, and other brands of pipetters.
- Tips are RNase- and DNase-free and ideal for handling RNA, PCR applications and working with radioisotopes or toxic chemicals
- Each tip lot is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, PCR, PCR and Real-Time PCR, RNA Extraction, Real Time PCR (qPCR), Total RNA Isolation, Total RNA from Animal Cells and Tissues, Transcriptome RNA Isolation, mRNA Isolation, microRNA Isolation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
100 μL | |
Sterile | |
Ambion™ | |
Store at room temperature. |
Filtered | |
Universal Fit | |
Compatible with Oxford Benchmnate/Nichiryo pipetters | |
Room Temperature |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.