Learn More
Ambion™ MessageAmp™ III RNA Amplification Kit Non-distribution product as customer accommodation.
Provides extensive improvements over existing methods: fewer components, pipetting steps, and manipulations in the workflow
Manufacturer:  Invitrogen 4383451
 View more versions of this product
Catalog No. 43-834-51
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Kit, RNA Amplification
The MessageAmp III aRNA Amplification Kit includes First-Strand Enzyme Mix, First-Strand Buffer Mix, Second-Strand Enzyme Mix, Second-Strand Buffer Mix, T7 Enzyme Mix, T7 Biotin IVT Mix, Control RNA (1mg/mL HeLa total RNA), Wash Buffer, and aRNA Binding Buffer, Filter Cartridges, Collection Tubes, 8-Strip PCR Tubes and Caps (0.2mL), 5X Array Fragmentation Buffer, and Nuclease-free Water
Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification
10 reactions