Ambion™ MessageAmp™ Premier RNA Amplification Kit Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Innovative kit for the preparation of RNA samples for microarray analysis
Manufacturer: Invitrogen 4383452
 View more versions of this product
Catalog No. 43-834-52
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Product Type Kit, RNA Amplification
    Includes 110μL First Strand Enzyme Mix, 440μL First Strand Buffer Mix, 220μL Second Strand Enzyme Mix, 550μL Second Strand Buffer Mix, 660μL T7 Enzyme Mix, 2 x 1.1mL T7 Biotin IVT Mix, 10μL Control RNA 1mg/mL, 1.75mL Nuclease-free Water, 6mL aRNA Binding Buffer Concentrate, 1.2mL RNA Binding Beads, 30mL aRNA Wash Solution Concentrate, 15mL aRNA Elution Solution, 1mL 5X Array Fragmentation Buffer, 2 U-Bottom Plates, 4 PCR Plates, 1 Reservoir, 10mL Nuclease-free Water
    For Use With (Application) Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification
    Quantity 100 reactions
  • Description & Specifications
Related Products
Product Content Feedback