Ambion™ MessageAmp™ Premier RNA Amplification Kit Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Innovative kit for the preparation of RNA samples for microarray analysis
Manufacturer: Invitrogen 4385821
 View more versions of this product
Catalog No. 43-858-21
  Check Availability
Add to Cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
    Specifications
    Product Type Kit, RNA Amplification
    Includes 11μL First Strand Enzyme Mix, 44μL First Strand Buffer Mix, 22μL Second Strand Enzyme Mix, 55μL Second Strand Buffer Mix, 66μL T7 Enzyme Mix, 220μL T7 Biotin IVT Mix, 10μL Control RNA 1mg/mL, 1.75mL Nuclease-free Water, 600μL aRNA Binding Buffer Concentrate, 120μL RNA Binding Beads, 10mL aRNA Wash Solution Concentrate, 5mL aRNA Elution Solution, 1mL 5X Array Fragmentation Buffer, 10 8-Strip PCR Tubes and Caps, 1 U-Bottom Plate, 1 Reservoir, 10mL Nuclease-free Water
    For Use With (Application) Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification
    Quantity 10 reactions
  • Description & Specifications
Related Products
Product Content Feedback