Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNAsecure™ Resuspension Solution (1 ml Tube)

Non-enzymatic reagent inactivate RNases in solution

Manufacturer:  Invitrogen™ AM7010

Encompass

Catalog No. AM7010


Add to cart

Description

Description

Invitrogen RNAsecure™ Resuspension Solution (patents pending) is a unique non-enzymatic reagent that will irreversibly inactivate RNases in solution. It is supplied at working concentration for direct resuspension of RNA pellets. Ten tubes containing 1mL each are provided. RNAsecure™ Resuspension Solution is specifically meant for resuspending RNA pellets that may contain trace amounts of ribonuclease (e.g., after total RNA isolation). Note: it cannot reverse RNA degradation that has already occurred, but can prevent further RNA degradation. To use, the RNA pellet is resuspended in the RNAsecure Resuspension Solution and heated to 60°C for 10 minutes It can be reheated to eliminate any RNase contamination introduced at a later time.

  • No post-treatment autoclaving
  • Safe, easy-to-use reagent inactivates RNases in solution
  • Inactivation process can be repeated to protect against newly introduced contaminants
  • Compatible with downstream procedures, such as RT-PCR and in vitro transcription

DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, PCR and Real-Time PCR, RNA Extraction, Real Time PCR (qPCR), Total RNA Isolation, Total RNA from Animal Cells and Tissues, mRNA Isolation

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

Store pure solution at –20°C.
Tubes
RNase Control Reagent
Ambion™, RNAsecure
Liquid
Molecular Biology Grade
1 mL
Dry Ice
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.