Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIzol™ Max™ Bacterial RNA Isolation Kit

Catalog No. p-4918749
Encompass_Preferred
Click to view available options
Quantity:
100 Preps
200 Preps

The TRIzol™ Max™ Bacterial RNA Isolation Kit provides a simple and reliable method to improve the isolation of intact total RNA from gram-positive and gram-negative bacteria.

The TRIzol™ Max™ Bacterial RNA Isolation Kit provides a simple and reliable method to improve the isolation of intact total RNA from gram-positive and gram-negative bacteria. The kit utilizes both the Max™ Bacterial Enhancement Reagent and TRIzol™ Reagent to inactivate endogenous RNases and promote protein denaturing, improving RNA quality and integrity. Isolation of RNA takes about an hour.

Key Features of the TRIzol™ Max™ Bacterial RNA Isolation Kit:
• Formulated for the isolation of RNA from both Gram-positive and Gram-negative bacteria
• Uses two technologies for maximum RNA yield and purity
• Minimizes time required for RNA isolation by eliminating enzymatic and mechanical lytic steps

Two Products in One Kit
The TRIzol™ Max™ Bacterial RNA Isolation Kit combines Max™ Bacterial Enhancement Reagent with TRIzol™ Reagent. A 5-minute pre-treatment of bacteria with Max™ Bacterial Enhancement Reagent, containing chelating agents, detergent and buffer, denatures bacterial proteins and effectively deactivates endogenous RNases. Subsequent bacterial lysis by TRIzol™ Reagent generates a high-quality, minimally degraded RNA product.

Purified RNA is Ideal for Use with a Variety of Applications and Products
Bacterial RNA isolated using the TRIzol™ Max™ Bacterial RNA Isolation Kit is suitable for downstream applications such as real-time PCR (qPCR), RT-PCR, northern blotting, nuclease protection assays, cDNA synthesis, microarray analysis, and dot blot hybridization.

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 20 mL Max Bacterial Enhancement Reagent; room temperature
• 100 mL TRIzol Reagent; room temperature

Isolation Technology Organic Extraction
Purification Time 1 hr.
Elution Volume 50 μL
Sample Type Bacteria (Gram +/-)
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling
Quantity 100 Preps
Shipping Condition Room Temperature
Starting Material Amount Cells: ≤108
Bacterial culture: 1.5 mL
Yield Gram-negative bacteria: >20 μg
Gram-positive bacteria: ≤3 μg
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.