Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ UltraPure™ Buffer-Saturated Phenol
UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids. The reagent, which consists of UltraPure™ Phenol that has been saturated with Tris-HCl buffer, is already buffer equilibrated to pH >7.4.
Supplier: Invitrogen™ 15513039
Description
- UltraPure™ Buffer-Saturated Phenol is used in the purification of nucleic acids
- The reagent, which consists of UltraPure™ Phenol that has been saturated with Tris-HCl buffer, is already buffer equilibrated to pH >7.4
- When mixtures are extracted with UltraPure™ Buffer- Saturated Phenol, proteins are denatured and collect in the organic phase or at the interphase, while most nucleic acids remain in the aqueous phase
- UltraPure™ Buffer-Saturated Phenol contains no preservatives
- It is packaged under an inert gas in shatter-resistant, plastic-coated amber bottles
- The appearance of the solution is evaluated at room temperature
- No RNase or DNase activity detected
- Store in refrigerator at 2°C to 8°C
Order Info
Shipping Condition: Room temperature
Specifications
Phenols | |
Bottle | |
UltraPure™ | |
100 mL | |
Liquid |
Store in refrigerator (2–8°C). | |
7.4 | |
Molecular Biology Grade | |
Room Temperature |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Provide Content Correction