Learn More
Invitrogen™ Urea
This high-purity molecular biology grade Invitrogen urea is ideal for the preparation of denaturing polyacrylamide gels used in nuclease protection assays.
Supplier: Invitrogen™ AM9902
Description
This high-purity molecular biology grade Invitrogen urea is ideal for the preparation of denaturing polyacrylamide gels used in nuclease protection assays. Supplied in one bottle containing 1 kg. Urea is thoroughly tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature
Specifications
Store at room temperature. | |
Bottle | |
Ambion™ | |
1000 g | |
Powder |
Nucleic Acid Gel Electrophoresis & Blotting | |
Urea | |
Molecular Biology Grade | |
Room Temperature |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.