Learn More
Ambion™ WT Expression Kit, High Throughput Non-distribution product as customer accommodation.
Generates amplified sense-strand cDNA ready for fragmentation and labeling using the Affymetrix™ GeneChip™ WT Terminal Labeling Kit
Manufacturer:  Invitrogen 4440536
 View more versions of this product
Catalog No. 44-405-36
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Kit, WT Expression
150μL First-Strand Enzyme Mix, 600μL First-Strand Buffer Mix, 650μL Second-Strand Enzyme Mix, 1.45mL Second-Strand Buffer Mix, 850μL IVT Enzyme Mix, 1.4mL IVT Buffer Mix, 10μL Control RNA 1mg/mL, 1.45mL 2nd-Cycle Buffer Mix, 300μL Random Primers, 1mL 2nd-Cycle Enzyme Mix, 300μL RNase H, 12.5mL Nucleic Acid Binding Buffer Concentrate, 2.5mL Nucleic Acid Binding Beads, 50mL Nucleic Acid Wash Solution Concentrate, 22mL Elution Solution
Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification
96 reactions