Learn More
Ambion™ WT Expression Kit, High Throughput Non-distribution product as customer accommodation.
Generates amplified sense-strand cDNA ready for fragmentation and labeling using the Affymetrix™ GeneChip™ WT Terminal Labeling Kit
Manufacturer:  Invitrogen 4440537
 View more versions of this product
Catalog No. 44-405-37
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Kit, WT Expression
50μL First-Strand Enzyme Mix, 160μL First-Strand Buffer Mix, 175μL Second-Strand Enzyme Mix, 400μL Second-Strand Buffer Mix, 210μL IVT Enzyme Mix, 750μL IVT Buffer Mix, 10μL Control RNA 1mg/mL, 425μL 2nd-Cycle Buffer Mix, 100μL Random Primers, 275μL 2nd-Cycle Enzyme Mix, 100μL RNase H, 3.5mL Nucleic Acid Binding Buffer Concentrate, 700μL Nucleic Acid Binding Beads, 15mL Nucleic Acid Wash Solution Concentrate, 6mL Elution Solution
Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification
24 reactions