Learn More
Ambion™ WT Expression Kit Non-distribution product as customer accommodation.
Designed to generate amplified sense-strand cDNA ready for fragmentation and labeling using the Affymetrix™ GeneChip™ WT Terminal Labeling Kit
Manufacturer:  Invitrogen 4411974
 View more versions of this product
Catalog No. 44-119-74
  Check Availability
Add to cart
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Kit, WT Expression
33μL First-Strand Enzyme Mix, 132μL First-Strand Buffer Mix, 165μL Second-Strand Enzyme Mix, 413μL Second-Strand Buffer Mix, 198μL IVT Enzyme Mix, 792μL IVT Buffer Mix, 10μL Control RNA 1mg/mL, 1.75mL Nuclease-free Water, 264μL 2nd-Cycle Buffer Mix, 66μL Random Primers, 264μL 2nd-Cycle Enzyme Mix, 66μL RNase H, 3.3mL Nucleic Acid Binding Buffer Concentrate, 660μL Nucleic Acid Binding Beads, 20mL Nucleic Acid Wash Solution Concentrate, 5mL Elution Solution, 10mL Nuclease-free Water, 40 8-Strip PCR Tubes and Caps, 4 U-Bottom Plate, 2 Reservoir
Expression Array Labeling, Gene Expression Analysis and Genotyping, Microarray Analysis, RNA Amplification
30 reactions