Learn More
Ambu™ MediBag™ Resuscitator and Rapid Drug Delivery System
The only resuscitator with built-in Metered Dose Inhaler (MDI) port and syringe medication port
Manufacturer:  Ambu, Inc. 461600000
 View more versions of this product
Catalog No. 19-065-687
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
6 Cs.
2.6L
Paediatric open reservoir tube
0.45L/2.6L
87.91 oz.
Built in metered dose inhaler port and syringe medication port