Learn More
Ambu™ Perfit ACE™ Extrication Collars Non-distribution product as customer accommodation.
Ambu Perfit ACE combined with the Mini Perfit ACE offer an incredible 28 sizes that ensure patients can have an effective accurate fit
Manufacturer:  Ambu™ 000281106
 View more versions of this product
Catalog No. 19-065-714
Add to cart
Description
Description
Features:
Sixteen-setting adjustability
Radiolucent, CT- and MRI-compatible plastic
Folds flat for easy storage
Two safety buttons lock adjustment
Flip chin-piece, easy access to airway
Horizontal sizing line and molded-in nasal cannula holders
Choose From:
Standard white collar
Olive drab military collar
Standard collar with two-strap waterproof Head Wedge™ immobilizer
flat-folding and radiolucent features of the extrication collar
strong adhesive attachment, compatible with all sizes of backboards
Also Available:
Mini Perfit ACE collars and Head Wedge immobilizers for infants, pediatrics, geriatrics, and small adults with 12 adjustable settings
Ambu Perfit Ace (19-170-404 and 19-170-407)
  • An adjustable collar designed to fit any adult with 16 different settings ranging from Neckless to Tall sizes
  • Incorporates latches that allow the rescuer to size the collar exactly to the victim's neck size
  • Two safety lock buttons hold the collar in place when adjusted
Ambu Mini Perfit ACE (19-065-714 and 19-065-715)
  • An adjustable collar designed to fit children and small adults
  • Has 12 different settings to provide an accurate fit
Specifications
Specifications
Yellow/light gray
Radiolucent, CT- and MRI-compatible plastic
3.5 oz.
Mini Perfit ACE collar only
Infant, paediatric and small adult radiolucent, CT and MRI
Infant, Pediatric and Small Adult
0.1kg