Ambu™ Perfit ACE™ Extrication Collars

Ambu Perfit ACE combined with the Mini Perfit ACE offer an incredible 28 sizes that ensure patients can have an effective accurate fit

Manufacturer: Ambu, Inc. 000264035

 View more versions of this product

Catalog No. 19-065-717

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) Infant, Pediatric and Adults radiolucent, CT and MRI
    Color Yellow
    Product Type Head wedge only
    Material Radiolucent, CT- and MRI-compatible plastic
    Size Infant, Pediatric and Adults

    Features:
    Sixteen-setting adjustability
    Radiolucent, CT- and MRI-compatible plastic
    Folds flat for easy storage
    Two safety buttons lock adjustment
    Flip chin-piece, easy access to airway
    Horizontal sizing line and molded-in nasal cannula holders
    Choose From:
    Standard white collar
    Olive drab military collar
    Standard collar with two-strap waterproof Head Wedge™ immobilizer
    flat-folding and radiolucent features of the extrication collar
    strong adhesive attachment, compatible with all sizes of backboards
    Also Available:
    Mini Perfit ACE collars and Head Wedge immobilizers for infants, pediatrics, geriatrics, and small adults with 12 adjustable settings

    Ambu Perfit Ace (19-170-404 and 19-170-407)

    • An adjustable collar designed to fit any adult with 16 different settings ranging from Neckless to Tall sizes
    • Incorporates latches that allow the rescuer to size the collar exactly to the victim's neck size
    • Two safety lock buttons hold the collar in place when adjusted
    Ambu Mini Perfit ACE (19-065-714 and 19-065-715)
    • An adjustable collar designed to fit children and small adults
    • Has 12 different settings to provide an accurate fit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit