Ambu Res-Cue Key™

Transparent plastic allows the victim's response to be seen

Manufacturer: Ambu, Inc. 248 201 103

 View more versions of this product

Catalog No. 18-999-297B

  Check Availability
Add to Cart

    Specifications

    Case Material Nylon
    Color Blue
    Includes Card sized instruction guide and basic CPR techniques
    Material Polyethylene
    Weight (English) 0.35 oz.
    Weight (Metric) 0.01kg
    For Use With (Application) Emergency resuscitation techniques, mouth-to-mouth, CPR procedures
    Product Type Res-Cue Key, Standard Woven Pack

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit