Ambu Res-Cue Key™

Transparent plastic allows the victim's response to be seen

Manufacturer: Ambu, Inc. 248 201 105

 View all options for this product

Catalog No. 18-999-297E


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Case Material Nylon
    Color Yellow
    Includes Card sized instruction guide and basic CPR techniques
    For Use With (Application) Emergency resuscitation techniques, mouth-to-mouth, CPR procedures
    Material Polyethylene
    Product Type Res-Cue Key, Standard Woven Pack
    Weight (English) 0.35 oz.
    Weight (Metric) 0.01kg

    Transparent plastic allows the victim's response to be seen
    Easy-to-use and inexpensive barrier that prevents mouth-to-mouth contact and the possibility of cross contamination. Includes a card-sized instruction guide and basic CPR techniques. Available in six colors.
    ·    Suitable for both adults and children
    ·    Elastic ear bands fit over victim's ears to keep mask in place
    ·    Allows standard mouth-to-mouth and mouth-to-nose techniques to be used without modification
    ·    One-way valve prevents contamination of the outer surface of mask

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit